Copper(I) bromide 98%

Supplier: Thermo Scientific Chemicals
Warning

11605-30 11605-A1
CAAA11605-30EA 156.26 CAD
CAAA11605-30 CAAA11605-A1
Copper(I) bromide 98%
Copper (I) bromide

Catalyst


Purity: 98% CAS No.: 7787-70-4 Molecular Formula: BrCu Formula Weight: 143.45 Physical Form: Powder Density: 4.98 Appearance: Pale green Melting Point: 492° Boiling Point: 1345° Solubility: Slightly soluble in cold water. Decomposes in hot water. Soluble in HBr, HCl, NH4OH Merck Reference: 14,2659 UN No.: 3077 MDL No.: MFCD00010969


Catalyst

Formula: BrCu
MW: 143.45 g/mol
Boiling Pt: 1345 °C
Melting Pt: 492 °C
Density: 4.98
Storage Temperature: Ambient
MDL Number: MFCD00010969
CAS Number: 7787-70-4
EINECS: 232-131-6
UN: 3077
ADR: 9,III
Merck Index: 14,2659

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR