Potassium iodate 99.4-100.4% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
11602-22
11602-36
CAAA11602-22EA
100.33
CAD
CAAA11602-22
CAAA11602-36
Potassium iodate 99.4-100.4% ACS
Potassium iodate
MDL: MFCD00011406
Slightly soluble in boiling water. Insoluble in alcohol.
Purity: 99.4-100.4%
CAS No.: 7758-05-6
Molecular Formula: KIO3
Formula Weight: 214.00
Physical Form: Powder
Density: 3.89
Melting Point: 560° dec.
Solubility: Slightly soluble in boiling water. Insoluble in alcohol
Merck Reference: 14,7642
UN No.: 1479
MDL No.: MFCD00011406
Formula:
KlO₃ Melting Pt: 560 °C (dec.) Density: 3.89 Storage Temperature: Ambient |
MDL Number:
MFCD00011406 CAS Number: 7758-05-6 EINECS: 231-831-9 UN: 1479 ADR: 5.1,II Merck Index: 14,7642 |
Specification Test Results
Insoluble matter | 0.005% |
pH of a 5 % solution | 5.0-8.0 at 25° |
Chloride and bromide (as Cl) | 0.01% |
I | 0.00001 |
Nitrogen compounds (as N) | 0.005% |
SO3 | 0.005% |
Heavy Metals (as Pb) | 5 ppm max. |
Fe | 0.001% |
NA | 0.005%% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...