Ammonium acetate ≥97.0% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Acetic acid ammonium salt
Reagent in analytical chemistry, drugs, textile dying, preserving meats, foam rubbers, vinyl plastics
Purity: 97.0% min
CAS No.: 631-61-8
Molecular Formula: CH3COONH4
Formula Weight: 77.08
Flash Point: 136°(277°F)
Physical Form: Crystalline
Density: 1.17
Sensitivity: Hygroscopic
Appearance: White
Odor: Ammonia-like
Melting Point: ca 114° dec.
Solubility: Soluble in cold water, approximately 148g/100ml. Decomposes in hot water. Soluble in methanol approximately 7.9g/100ml
Beilstein Reference: 4186741
Merck Reference: 14,495
MDL No.: MFCD00013066
Reagent in analytical chemistry, drugs, textile dying, preserving meats, foam rubbers, vinyl plastics
Formula:
H₃CCOONH₄ Melting Pt: ∼114 °C (dec.) Density: 1.17 Flash Pt: 136 °C (277 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00013066 CAS Number: 631-61-8 EINECS: 211-162-9 Merck Index: 14,495 |
|
Specification Test Results
Insoluble matter | 0.0005% |
Residue after ignition | 0.01% |
pH of a 5% solution | 6.7-7.3 at 25° |
Cl | 5 ppm |
NO3 | 0.001% |
SO3 | 0.001% |
Heavy Metals (as Pb) | 5 ppm max. |
Fe | 5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...