Ammonium dihydrogen phosphate ≥98.0% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
11598-36
11598-A3
CAAA11598-36EA
112.02
CAD
CAAA11598-36
CAAA11598-A3
Ammonium dihydrogen phosphate ≥98.0% ACS
Ammonium dihydrogen phosphate
Preparation of baking powder with sodium bicarbonate, in fermentation
Purity: 98.0% min
CAS No.: 7722-76-1
Molecular Formula: NH4H2PO4
Formula Weight: 115.03
Physical Form: Crystalline
Density: 1.803
Melting Point: 190°
Solubility: Freely soluble in water. Slightly soluble in alcohol. Insoluble in acetone
Merck Reference: 14,543
MDL No.: MFCD00003396
Preparation of baking powder with sodium bicarbonate, in fermentation
Formula:
NH₄H₂PO₄ Melting Pt: 190 °C Density: 1.803 Storage Temperature: Ambient |
MDL Number:
MFCD00003396 CAS Number: 7722-76-1 EINECS: 231-764-5 Merck Index: 14,543 |
Specification Test Results
Insoluble matter | 0.005% |
pH of a 5 % solution | 3.8-4.4 at 25° |
Cl | 5 ppm |
NO3 | 0.00001 |
SO3 | 0.01% |
Heavy Metals (as Pb) | 5% |
Fe | 0.001% |
Ca | 0.001% |
MG | 0.0005% |
K | 0.005% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...