Diammonium hydrogen orthophosphate ≥98.0% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Diammonium hydrogen orthophosphate, Ammonium hydrogen phosphate, di-Ammonium hydrogen phosphate, Ammonium Phosphate, Ammonium biphosphate, Fyrex, Ammonium phosphate dibasic, Ammonium monohydrogen phosphate, Diammonium hydrogen phosphate
MDL: MFCD00010891
Purity: 98.0% min
CAS No.: 7783-28-0
Molecular Formula: (NH4)2HPO4
Formula Weight: 132.06
Physical Form: Crystalline
Density: 1.619
Appearance: White
Melting Point: 155° dec.
Merck Reference: 14,542
MDL No.: MFCD00010891
Formula:
(NH₄)₂HPO₄ MW: 132.06 g/mol Melting Pt: 155 °C (dec.) Density: 1.619 Storage Temperature: Ambient |
MDL Number:
MFCD00010891 CAS Number: 7783-28-0 EINECS: 231-987-8 REACH: 01-2119490974-22 Merck Index: 14,542 |
Specification Test Results
Insoluble matter | 0.005% |
pH of a 5 % solution | 7.7-8.1 at 25° |
Cl | 0.001% |
NO3 | 0.003% |
SO3 | 0.01% |
Heavy Metals (as Pb) | 0.001% |
Fe | 0.001% |
Ca | 0.001% |
MG | 0.0005% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...