Magnesium sulfate heptahydrate 98.0-102.0% ACS

Supplier: Thermo Scientific Chemicals

Synonyms: sulphuric acid magnesium salt heptahydrate, sulfuric acid magnesium salt heptahydrate, Epsom salt, Bitter salt

11596-22 11596-36 11596-A3
CAAA11596-22EA 58.93 CAD
CAAA11596-22 CAAA11596-36 CAAA11596-A3
Magnesium sulfate heptahydrate 98.0-102.0% ACS
Magnesium sulphate heptahydrate

Fireproofing, textiles, catalyst carrier, ceramics


Purity: 98.0-102.0% CAS No.: 10034-99-8 Molecular Formula: MgSO4.7H2O Formula Weight: 246.48 (120.37anhy) Physical Form: Crystalline Density: 1.68 Solubility: Soluble in water. Slightly soluble in alcohol, glycerol Merck Reference: 14,5691 MDL No.: MFCD00149785


Fireproofing, textiles, catalyst carrier, ceramics

Formula: MgSO₄·7H₂O
MW: 246.48 g/mol
Melting Pt: 1124 °C
Density: 1.68
Storage Temperature: Ambient
MDL Number: MFCD00149785
CAS Number: 10034-99-8
EINECS: 231-298-2
Merck Index: 14,5691

Order Now

Specification Test Results

Insoluble matter 0.005%
pH of a 5 solution 5.0-8.2 at 25°
Cl 5 ppm
NO3 0.002%
NH3 0.002%
Ca 0.02%
Heavy Metals (as Pb) 5 ppm
Fe 5 ppm
Mn 5 ppm
K 0.00005

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR