Potassium chloride 99.0-100.5% ACS
Supplier: Thermo Scientific Chemicals
Warning
|
In buffer solution, electrode cells, photography, lab reagent, fertilizer, spectroscopy
Purity: 99.0-100.5%
CAS No.: 7447-40-7
Molecular Formula: KCl
Formula Weight: 74.55
Physical Form: Crystalline
Density: 1.984
Refractive Index: 1.490
Sensitivity: Hygroscopic
Odor: Odorless
Melting Point: 773°
Boiling Point: 1500° subl.
Solubility: Freely soluble in water. Soluble in glycerol, alcohol
Merck Reference: 14,7621
MDL No.: MFCD00011360
In buffer solution, electrode cells, photography, lab reagent, fertilizer, spectroscopy
Formula:
KCl Boiling Pt: 1500 °C (subl.) Melting Pt: 773 °C Density: 1.984 Storage Temperature: Ambient |
MDL Number:
MFCD00011360 CAS Number: 7447-40-7 EINECS: 231-211-8 Merck Index: 14,7621 |
Specification Test Results
Insoluble matter | 0.005% |
pH of a 5 % solution | 5.4-8.6 at 25° |
I | 0.002% |
Br | 0.01% |
NO3 | 0.003% |
PO3 | 5 ppm |
SO3 | 0.001% |
Ba | Pass test |
Ca | 0.002%% |
MG | 0.001% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...