Disodium hydrogen phosphate heptahydrate 98.0-102.0% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
11592-36
11592-A3
CAAA11592-36EA
122.45
CAD
CAAA11592-36
CAAA11592-A3
Disodium hydrogen phosphate heptahydrate 98.0-102.0% ACS
di-Sodium hydrogen phosphate heptahydrate
MDL: MFCD00149180
Purity: 98.0-102.0%
CAS No.: 7782-85-6
Molecular Formula: Na2HPO4.7H2O
Formula Weight: 268.07 (141.98anhy)
Physical Form: Crystalline
Density: 1.7
Appearance: White
Odor: Odorless
Melting Point: 48.1°-5H2O
Merck Reference: 14,8659
MDL No.: MFCD00149180
Formula:
HNa₂O₄P·7H₂O Melting Pt: 48.1 °C (-5H₂O) Density: 1.7 Storage Temperature: Ambient |
MDL Number:
MFCD00149180 CAS Number: 7782-85-6 EINECS: 231-448-7 Merck Index: 14,8659 |
Specification Test Results
pH of a 5% solution at 25° | 8.7-9.3 at 25° |
Insoluble matter | 0.005% |
Cl | 0.001% |
SO3 | 0.005% |
Heavy Metals (as Pb) | 0.001% |
Fe | 0.001% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...