Sodium dihydrogen phosphate monohydrate 98,0-102,0 ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sodium phosphate monobasic monohydrate, Sodium dihydrogen orthophosphate monohydrate, Monosodium phosphate monohydrate, Sodium dihydrogenorthophosphate monohydrate, Sodium orthophosphate monobasic monohydrate

11591-30 11591-A1 11591-A7
CAAA11591-30EA 91.48 CAD
CAAA11591-30 CAAA11591-A1 CAAA11591-A7
Sodium dihydrogen phosphate monohydrate 98,0-102,0 ACS
Sodium dihydrogen phosphate monohydrate

MDL: MFCD00149208 Fieser: 1,395


Purity: 98.0-102.0% CAS No.: 10049-21-5 Molecular Formula: NaH2PO4.H2O Formula Weight: 137.99 (119.98anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Merck Reference: 14,8660 MDL No.: MFCD00149208

Formula: NaH₂PO₄·H₂O
MW: 137.99 g/mol
Melting Pt: 100
Density: 2.04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149208
CAS Number: 10049-21-5
EINECS: 231-449-2
Merck Index: 14,8660
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4e3b73d9200cc18a9423c5b98ad4affbd4aca8a&vid=a46f80a2f4b075e731486341a7be9b01c444a160&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Insoluble matter 0.01%
pH of a 5 solution 4.1-4.5 at 25
Cl 5 ppm
SO3 0.003%
Ca 0.005%
K 0.01%
As 0.5 ppm
Heavy Metals (as Pb) 0.00001
Fe 0.001%%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR