Antimony(III) oxide ≥99.6%

Supplier: Thermo Scientific Chemicals
Warning

11579-18 11579-36 11579-A3
CAAA11579-18EA 36.81 CAD
CAAA11579-18 CAAA11579-36 CAAA11579-A3
Antimony(III) oxide ≥99.6%
Antimony (III) oxide

MDL: MFCD00011214 Slightly soluble in water, Slowly dissolves in warm HCl or warm KOH solutions


Purity: 99.6% min CAS No.: 1309-64-4 Molecular Formula: Sb2O3 Formula Weight: 291.50 Physical Form: 1.1-1.8 Micron Powder Density: 5.67 Appearance: White Odor: Odorless Melting Point: 656° Boiling Point: 1550° subl. Solubility: Slightly soluble in water, Slowly dissolves in warm HCl or warm KOH solutions Merck Reference: 14,711 MDL No.: MFCD00011214

Formula: O₃Sb₂
Boiling Pt: 1550 °C (subl.)
Melting Pt: 656 °C
Density: 5.67
Storage Temperature: Ambient
MDL Number: MFCD00011214
CAS Number: 1309-64-4
EINECS: 215-175-0
UN: 1549
ADR: 6.1,III
Merck Index: 14,711

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR