Tin(IV) chloride, anhydrous ≥98% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
MDL: MFCD00011242
Soluble in alcohol, CCl4, benzene, toluene, and acetone. Soluble in water with much heat evolution.
Fieser: 1,1111 14,304 15,311 17,335 18,354 19,338 20,373 21,422
Purity: 98%
CAS No.: 7646-78-8
Molecular Formula: Cl4Sn
Formula Weight: 260.50
Physical Form: Liquid
Density: 2.26
Refractive Index: 1.512
Sensitivity: Moisture Sensitive
Appearance: Colorless
Melting Point: -33°
Boiling Point: 114°
Solubility: Soluble in alcohol, CCl4, benzene, toluene, and acetone. Soluble in water with much heat evolution
Merck Reference: 14,8774
UN No.: 1827
MDL No.: MFCD00011242
Formula:
Cl₄Sn Boiling Pt: 114 °C Melting Pt: –33 °C Density: 2.26 Storage Temperature: Ambient |
MDL Number:
MFCD00011242 CAS Number: 7646-78-8 EINECS: 231-588-9 UN: 1827 ADR: 8,II Merck Index: 14,8774 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...