Tin(IV) chloride pentahydrate 98%, lumps

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tin tetrachloride pentahydrate, Stannic chloride pentahydrate

11570-22 11570-36
CAAA11570-22EA 63.36 CAD
CAAA11570-22 CAAA11570-36
Tin(IV) chloride pentahydrate 98%, lumps
Tin (IV) chloride pentahydrate

Tin(IV) chloride hydrate is used in the formation of a xerogel via a cyanogel, a gel that has potential use in solid-state gas sensors. It is involved in the preparation of Indium tin oxide thin films by chemical vapor deposition method.


Purity: 98% CAS No.: 7646-78-8 Molecular Formula: Cl4Sn.xH2O Formula Weight: 260.50(anhy) Physical Form: Lump Appearance: Off-white Odor: Pungent Melting Point: 56° Solubility: Soluble in water, alcohol Merck Reference: 14,8774 UN No.: 2440 MDL No.: MFCD00149864

Formula: Cl₄Sn·5H₂O
MW: 350.6 g/mol
Melting Pt: 56 °C
Density: 2.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149864
CAS Number: 10026-06-9
EINECS: 231-588-9
UN: 2440
ADR: 8,III
Merck Index: 14,8774

Order Now

Specification Test Results

Assay (unspecified) 97.6% min

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR