Ammonium fluoride ≥98.0% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
11568-14
11568-30
11568-A1
11568-D9
CAAA11568-14EA
109.34
CAD
CAAA11568-14
CAAA11568-30
CAAA11568-A1
CAAA11568-D9
Ammonium fluoride ≥98.0% ACS
Ammonium fluoride
For etching glass, in printing textile, for preservation of wood
Purity: 98.0% min
CAS No.: 12125-01-8
Molecular Formula: NH4F
Formula Weight: 37.04
Physical Form: Crystalline
Density: 1.01
Solubility: Soluble in cold water (0°C). Decomposes in hot water to NH3 and ammonium bifluoride. Slightly soluble in alcohol
Merck Reference: 14,522
UN No.: 2505
MDL No.: MFCD00011423
For etching glass, in printing textile, for preservation of wood
Formula:
NH₄F Melting Pt: 160 °C Density: 1.01 Storage Temperature: Ambient |
MDL Number:
MFCD00011423 CAS Number: 12125-01-8 EINECS: 235-185-9 UN: 2505 ADR: 6.1,III Merck Index: 14,522 |
Specification Test Results
Insoluble matter | 0.005% |
Residue after ignition | 0.01% |
Cl | 0.001% |
SO3 | 0.005% |
Heavy Metals (as Pb) | 5 ppm max. |
Fe | 5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...