Chromium(III) nitrate nonahydrate ≥98.5%

Supplier: Thermo Scientific Chemicals
Danger

11565-14 11565-30 11565-A1
CAAA11565-14EA 70.63 CAD
CAAA11565-14 CAAA11565-30 CAAA11565-A1
Chromium(III) nitrate nonahydrate ≥98.5%
Chromium (III) nitrate nonahydrate

Catalyst, corrosion inhibitor, in textile printing


Purity: 98.5% CAS No.: 7789-02-8 Molecular Formula: Cr(NO3)3.9H2O Formula Weight: 400.14 (238.01anhy) Physical Form: Crystalline Appearance: Purple Odor: Odorless Melting Point: 60° Solubility: Soluble in water, alcohol Merck Reference: 14,2226 UN No.: 2720 MDL No.: MFCD00149659


Catalyst, corrosion inhibitor, in textile printing

Formula: CrN₃O₉·9H₂O
Melting Pt: 60 °C
Density: 1.807 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149659
CAS Number: 7789-02-8
EINECS: 236-921-1
UN: 2720
ADR: 5.1,III
Merck Index: 14,2226

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR