Magnesium nitrate hexahydrate 98.0-102.0% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
As fertilizer, in manufacture of ammonium nitrate, as a reagent chemical, and in pyrotechnics
Purity: 98.0-102.0%
CAS No.: 13446-18-9
Molecular Formula: Mg(NO3)2.6H2O
Formula Weight: 256.41 (148.32anhy)
Physical Form: Crystalline
Density: 1.46
Melting Point: 89°
Boiling Point: 330° dec.
Solubility: Soluble in water, alcohol and liquid NH3
Merck Reference: 14,5674
UN No.: 1474
MDL No.: MFCD00149779
As fertilizer, in manufacture of ammonium nitrate, as a reagent chemical, and in pyrotechnics
Formula:
Mg(NO₃)₂·6H₂O Boiling Pt: 330 °C (dec.) Melting Pt: 89 °C Density: 1.46 Flash Pt: 93.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149779 CAS Number: 13446-18-9 EINECS: 233-826-7 UN: 1474 ADR: 5.1,III Merck Index: 14,5674 |
Specification Test Results
Insoluble matter | 0.005% |
pH of a 5% solution | 5.0-8.2 at 25° |
Cl | 0.001% |
PO3 | 5 ppm |
SO3 | 0.005% |
NH3 | 0.003% |
Ba | 0.00005 |
Ca | 0.0001 |
Heavy Metals (as Pb) | 5 ppm |
Fe | 5ppm ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...