Manganese(II) chloride tetrahydrate ≥99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Manganese dichloride tetrahydrate, Manganese chloride (tetrahydrate), Manganese(II) chloride tetrahydrate, Manganous chloride tetrahydrate, Manganese chloride tetrahydrate, Manganese Chloride, Tetrahydrate

11563-22 11563-36
CAAA11563-22EA 67.94 CAD
CAAA11563-22 CAAA11563-36
Manganese(II) chloride tetrahydrate ≥99% (metals basis)
Manganese (II) chloride tetrahydrate

MDL: MFCD00149792


Purity: 99% CAS No.: 13446-34-9 Molecular Formula: Cl2Mn.4H2O Formula Weight: 197.90 (125.84anhy) Physical Form: Crystalline Density: 2.010 Appearance: Pink Odor: Odorless Melting Point: 58° Merck Reference: 14,5728 MDL No.: MFCD00149792

Formula: MnCl₂·4H₂O
MW: 197.9 g/mol
Boiling Pt: 1190 °C (1013 hPa)
Melting Pt: 58 °C
Density: 2.010
Storage Temperature: Ambient
MDL Number: MFCD00149792
CAS Number: 13446-34-9
EINECS: 231-869-6
UN: 3077
ADR: 9,III
Merck Index: 14,5728

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR