Sodium tetrafluoroborate ≥95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
11562-36
11562-A3
CAAA11562-36EA
98.59
CAD
CAAA11562-36
CAAA11562-A3
Sodium tetrafluoroborate ≥95%
Sodium tetrafluoroborate
Fluorinating agent
Purity: 95%
CAS No.: 13755-29-8
Molecular Formula: BF4Na
Formula Weight: 109.79
Physical Form: Lump/Granular
Density: 2.47
Appearance: White to off-white
Melting Point: 384° dec.
Solubility: Soluble in water
Merck Reference: 14,8617
MDL No.: MFCD00003515
Fluorinating agent
Formula:
BF₄Na Melting Pt: 384 °C (dec.) Density: 2.47 Storage Temperature: Ambient |
MDL Number:
MFCD00003515 CAS Number: 13755-29-8 EINECS: 237-340-6 Merck Index: 14,8617 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...