Sodium fluoride ≥99% ACS

Supplier: Thermo Scientific Chemicals
Danger

11561-30 11561-A1
CAAA11561-30EA 110.76 CAD
CAAA11561-30 CAAA11561-A1
Sodium fluoride ≥99% ACS
Sodium fluoride

Steel degreasing agent, in tin electroplating, flux, heat treating salt compositions, fluoridation of drinking water, removing HF from exhaust systems to reduce air pollution, frosting glasses


Purity: 99% min CAS No.: 7681-49-4 Molecular Formula: NaF Formula Weight: 41.99 Physical Form: Powder Density: 2.78 Refractive Index: 1.336 Odor: Odorless Melting Point: 993° Boiling Point: 1704° Solubility: Soluble in water. Insoluble in alcohol Merck Reference: 14,8618 UN No.: 1690 MDL No.: MFCD00003524


Steel degreasing agent, in tin electroplating, flux, heat treating salt compositions, fluoridation of drinking water, removing HF from exhaust systems to reduce air pollution, frosting glasses

Formula: NaF
Boiling Pt: 1704 °C
Melting Pt: 993 °C
Density: 2.78
Storage Temperature: Ambient
MDL Number: MFCD00003524
CAS Number: 7681-49-4
EINECS: 231-667-8
UN: 1690
ADR: 6.1,III
Merck Index: 14,8618
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2807d129a60e4d136eb782386d584317338c4499&vid=02396e9a739aa8e35f0b20961710e28c6ec76be0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Insoluble matter 0.02%
Loss on drying at 150°C 0.3%
Cl 0.005%
Titratable acid 0.03meq/g
Titratable base 0.01meq/g
Na2SiF3 0.1%
SO3 0.03%
PO3 0.005%
Heavy Metals (as Pb) 0.003%%
Fe 0.003%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR