Sodium sulfate ≥99.0%, powder ACS

Supplier: Thermo Scientific Chemicals

Synonyms: Disodium sulfate, Bisodium sulfate, Disodium sulphate, Sulfuric acid disodium salt, Bisodium sulphate, Sulphuric acid disodium salt

11560-36 11560-A3 11560-A9
CAAA11560-36EA 91.8 CAD
CAAA11560-36 CAAA11560-A3 CAAA11560-A9
Sodium sulfate ≥99.0%, powder ACS
Sodium sulphate

In drying organic liquids, in Kjeldahl nitrogen determination, in manufacturing of glass. Sodium sulfate anhydrous is used primarily for drying non-aqueous solvents by removing traces of water. It is used as a fining agent for molten glass, levelling agent in textile processing, and filler in detergents. It is also used in Kraft process for paper pulping, defrosting windows, manufacture of starch, glass, and soap.


Notes: Anhydrous sodium sulfate is hygroscopic in nature, and absorbs moisture converting to the hydrated form. The hydrated form on heating gets converted back to anhydrous form.


In drying organic liquids, in Kjeldahl nitrogen determination, in manufacturing of glass

Formula: Na₂SO₄
MW: 142.04 g/mol
Boiling Pt: 1429
Melting Pt: 888
Density: 2.680
Storage Temperature: Ambient
MDL Number: MFCD00003504
CAS Number: 7757-82-6
EINECS: 231-820-9
Merck Index: 14,8680
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4977cb8ca54638f22a60675bfbef71bbb77ab6&vid=942501aaa5afc8d85b82628854aac94d517322eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Insoluble matter 0.01%
Loss on ignition 0.5%
Cl 0.001%
N 5 ppm
PO3 0.001%
Heavy Metals (as Pb) 5 ppm
Fe 0.001%
Ca 0.01%
MG 0.005%%
K 0.01%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR