Zinc acetate dihydrate 98.0-101.0% ACS

Supplier: Thermo Scientific Chemicals
Warning

11559-30 11559-A1 11559-A7
CAAA11559-30EA 58.93 CAD
CAAA11559-30 CAAA11559-A1 CAAA11559-A7
Zinc acetate dihydrate 98.0-101.0% ACS
Zinc acetate dihydrate

MDL: MFCD00066961 Beilstein Registry No.: 3732513 Soluble in water, alcohol


Purity: 98.0-101.0% CAS No.: 5970-45-6 Molecular Formula: C4H10O6Zn Formula Weight: 219.50 (183.47anhy) Physical Form: Crystalline Density: 1.735 Appearance: White Odor: Weak, characteristic pH: 6-7 Melting Point: 237° Solubility: Soluble in water, alcohol Beilstein Reference: 3732513 Merck Reference: 14,10128 UN No.: 3077 MDL No.: MFCD00066961

Formula: (H₃CCOO)₂Zn·2H₂O
Melting Pt: 237 °C
Density: 1.735
Storage Temperature: Ambient
MDL Number: MFCD00066961
CAS Number: 5970-45-6
EINECS: 209-170-2
UN: 3077
ADR: 9,III
Merck Index: 14,10128

Order Now

Specification Test Results

Insoluble matter 0.005%
pH of a 5% solution 6.0-7.0 at 25°
Cl 5 ppm
SO3 0.005%
Ca 0.005%
Fe 5 ppm
PB 0.002%
MG 0.00005
K 0.01%
NA 0.05%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR