Zinc oxide ≥99.0% ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Zinci oxidum, Zinc (II) oxide

11558-36 11558-A3
CAAA11558-36EA 113.13 CAD
CAAA11558-36 CAAA11558-A3
Zinc oxide ≥99.0% ACS
Zinc oxide

Pigment in white paints, UV absorber in plastics, ceramics, piezoelectric devices


Purity: 99.0% min CAS No.: 1314-13-2 Molecular Formula: ZnO Formula Weight: 81.37 Physical Form: -325 Mesh Powder Density: 5.606 Melting Point: 1975° Solubility: Insoluble in water. Soluble in dilute acetic acid, dilute mineral acids, ammonia, ammonium carbonate Merck Reference: 14,10147 UN No.: 3077 MDL No.: MFCD00011300


Pigment in white paints, UV absorber in plastics, ceramics, piezoelectric devices

Formula: ZnO
MW: 81.39 g/mol
Melting Pt: 1975 °C
Density: 5.606
Storage Temperature: Ambient
MDL Number: MFCD00011300
CAS Number: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9 III
REACH: 01-2119463881-32
Merck Index: 14,10147

Order Now

Specification Test Results

Insoluble in dilute sulfuric acid 0.01%
Alkalinity Pass test
Cl 0.001%
NO3 0.003%
Sulfur compounds (as SO3) 0.01%
Ca 0.005%
Fe 0.001%
PB 0.005%
MG 0.005%%
Mn 5ppm ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR