Potassium tetrafluoroborate ≥99% typ. >99.5%

Supplier: Thermo Scientific Chemicals
Warning

11556-36 11556-A3
CAAA11556-36EA 84.37 CAD
CAAA11556-36 CAAA11556-A3
Potassium tetrafluoroborate ≥99% typ. >99.5%
Potassium tetrafluoroborate

MDL: MFCD00011395 Soluble in water (0.45g/100g H2O). Slightly soluble in boiling alcohol


Purity: 99% min CAS No.: 14075-53-7 Molecular Formula: BF4K Formula Weight: 125.91 Physical Form: Powder Density: 2.50 Refractive Index: 1.3245 Sensitivity: Hygroscopic Melting Point: 529° Boiling Point: dec. Solubility: Soluble in water (0.45g/100g H2O). Slightly soluble in boiling alcohol Merck Reference: 14,7686 MDL No.: MFCD00011395

Formula: BF₄K
Boiling Pt: Dec.
Melting Pt: 529 °C
Density: 2.50
Storage Temperature: Ambient
MDL Number: MFCD00011395
CAS Number: 14075-53-7
EINECS: 237-928-2
Merck Index: 14,7686

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR