Potassium fluoride dihydrate ≥98.5%
Supplier: Thermo Scientific Chemicals
|
Danger
|
11555-14
11555-30
11555-A1
CAAA11555-14EA
53.09
CAD
CAAA11555-14
CAAA11555-30
CAAA11555-A1
Potassium fluoride dihydrate ≥98.5%
Potassium fluoride dihydrate
MDL: MFCD00149929
Freely soluble in water
Purity: 98.5+%
CAS No.: 13455-21-5
Molecular Formula: KF.2H2O
Formula Weight: 94.13 (58.10anhy)
Physical Form: Crystalline
Density: 2.5
Refractive Index: 1.352
Sensitivity: Hygroscopic
Odor: Odorless
Melting Point: 41°
Solubility: Freely soluble in water
Merck Reference: 14,7632
UN No.: 1812
MDL No.: MFCD00149929
Formula:
FK·2H₂O MW: 94.13 g/mol Boiling Pt: 1505 °C (1013 hPa) Melting Pt: 41 °C Density: 2.5 Storage Temperature: Ambient |
MDL Number:
MFCD00149929 CAS Number: 13455-21-5 EINECS: 232-151-5 UN: 1812 ADR: 6.1,III Merck Index: 14,7632 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...