Sodium acetate, anhydrous ≥99.0% ACS
Supplier: Thermo Scientific Chemicals
Warning
|
11554-30
11554-A1
11554-A7
CAAA11554-30EA
64.46
CAD
CAAA11554-30
CAAA11554-A1
CAAA11554-A7
Sodium acetate, anhydrous ≥99.0% ACS
Sodium acetate
MDL: MFCD00012459
Beilstein Registry No.: 3595639
Fieser: 1,1024 5,59
Purity: 99.0% min
CAS No.: 127-09-3
Molecular Formula: NaOOCCH3
Formula Weight: 82.03
Physical Form: Crystalline
Density: 1.528
pH: 7,5-9
Melting Point: 324°
Beilstein Reference: 3595639
Merck Reference: 14,8571
MDL No.: MFCD00012459
Formula:
H₃CCOONa Boiling Pt: 881.4 °C (1013 hPa) Melting Pt: 324 °C Density: 1.528 Flash Pt: >250 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00012459 CAS Number: 127-09-3 EINECS: 204-823-8 REACH: 01-2119485123-42 Merck Index: 14,8571 |
Specification Test Results
Insoluble matter | 0.01% |
Loss on drying at 120°C | 1% |
pH of a 5% solution | 7.5-9.2 at 25° |
Cl | 0.002% |
PO3 | 0.001% |
SO3 | 0.003% |
Ca | 0.005% |
MG | 0.002% |
Heavy Metals (as Pb) | 0.001%% |
Fe | 0.001% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...