Sodium acetate trihydrate 99-101% ACS
Supplier: Thermo Scientific Chemicals
Synonyms:
Acetic acid sodium salt trihydrate
Common Applications: In photography, analytical laboratory as buffer, pharmaceutical
Purity: 99.0%-100.5%
CAS No.: 6131-90-4
Molecular Formula: NaOOCCH3.3H2O
Formula Weight: 136.08 (82.03anhy)
Physical Form: Crystalline
Density: 1.45
pH: 7.5-9.0
Melting Point: 62-64°
Solubility: Freely soluble in alcohol
Beilstein Reference: 3732037
Merck Reference: 14,8571
MDL No.: MFCD00071557
In photography, analytical laboratory as buffer, pharmaceutical
Formula:
H₃CCOONa·3H₂O Boiling Pt: 120…123 °C (1013 hPa) Melting Pt: 62…64 °C Density: 1.45 Storage Temperature: Ambient |
MDL Number:
MFCD00071557 CAS Number: 6131-90-4 EINECS: 204-823-8 Merck Index: 14,8571 |
|
Specification Test Results
Insoluble matter | 0.005% |
pH of a 5% solution | 7.5-9.2 at 25° |
Cl | 0.001% |
PO3 | 5 ppm |
SO3 | 0.002% |
Ca | 0.005% |
MG | 0.002% |
Heavy Metals (as Pb) | 5 ppm |
Fe | 5 ppm |
Substances reducing permanganate P.T. | Pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...