Niobium(V) chloride ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Niobium pentachloride
11548-14
11548-22
11548-36
CAAA11548-14EA
150.42
CAD
CAAA11548-14
CAAA11548-22
CAAA11548-36
Niobium(V) chloride ≥99.9% (metals basis)
Niobium (V) chloride
MDL: MFCD00011127
Purity: 99.9%
CAS No.: 10026-12-7
Molecular Formula: Cl5Nb
Formula Weight: 270.20
Physical Form: -100 Mesh Powder, Packaged under argon
Density: 2.74
Appearance: Yellow
Odor: Acidic
Melting Point: 208.3°
Boiling Point: 248.2°
Merck Reference: 14,6557
UN No.: 3260
MDL No.: MFCD00011127
Formula:
Cl₅Nb MW: 270.17 g/mol Boiling Pt: 254 Melting Pt: 205 Density: 2.74 Storage Temperature: Ambient |
MDL Number:
MFCD00011127 CAS Number: 10026-12-7 EINECS: 233-059-8 UN: 3260 ADR: 8 III Merck Index: 14,6557 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...