Aluminium nitride N ≥ 32.0%

Supplier: Thermo Scientific Chemicals
Warning

11546-06 11546-18 11546-30 11546-A1
CAAA11546-06EA 62.09 CAD
CAAA11546-06 CAAA11546-18 CAAA11546-30 CAAA11546-A1
Aluminium nitride N ≥ 32.0%
Aluminium nitride

MDL: MFCD00003429 Notes: Decomposes in water


CAS No.: 24304-00-5 Molecular Formula: AlN Formula Weight: 40.99 Physical Form: -325 Mesh Powder, typically 4 Micron APS Density: 3.26 Sensitivity: Moisture Sensitive Appearance: White Odor: Ammonia-like Melting Point: 2200° Merck Reference: 14,353 MDL No.: MFCD00003429

Formula: AlN
Boiling Pt: 2517 °C (1013 hPa)
Melting Pt: >2200 °C
Density: 3.26
Storage Temperature: Ambient
MDL Number: MFCD00003429
CAS Number: 24304-00-5
EINECS: 246-140-8
Merck Index: 14,353

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR