Tin(II) fluoride ≥97.5%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Tin difluoride
11544-22
11544-36
CAAA11544-22EA
81.05
CAD
CAAA11544-22
CAAA11544-36
Tin(II) fluoride ≥97.5%
Tin (II) fluoride
MDL: MFCD00042540
Purity: 97.5%
CAS No.: 7783-47-3
Molecular Formula: F2Sn
Formula Weight: 156.69
Physical Form: Crystalline
Density: 4.57
Appearance: White to off-white
Odor: Odorless
Melting Point: 219°
Boiling Point: 850°
Merck Reference: 14,8784
UN No.: 3260
MDL No.: MFCD00042540
Formula:
SnF₂ MW: 156.71 g/mol Boiling Pt: 850 °C Melting Pt: 219 °C Density: 4.57 Storage Temperature: Ambient |
MDL Number:
MFCD00042540 CAS Number: 7783-47-3 EINECS: 231-999-3 UN: 3260 ADR: 8,II Merck Index: 14,8784 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...