Vanadium(IV) fluoride ≥95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
11543-06
11543-14
CAAA11543-06EA
92.11
CAD
CAAA11543-06
CAAA11543-14
Vanadium(IV) fluoride ≥95%
Vanadium (IV) fluoride
MDL: MFCD00042554
Soluble in water (blue color), acetone (deep green color), and glacial acetic acid (blue-green color)
Purity: 95%
CAS No.: 10049-16-8
Molecular Formula: F4V
Formula Weight: 126.94
Physical Form: Powder
Density: 2.975
Appearance: Yellow to brown
Melting Point: 325° dec.
Solubility: Soluble in water (blue color), acetone (deep green color), and glacial acetic acid (blue-green color)
UN No.: 2923
MDL No.: MFCD00042554
Formula:
F₄V Melting Pt: 325 °C (dec.) Density: 2.975 Storage Temperature: Ambient |
MDL Number:
MFCD00042554 CAS Number: 10049-16-8 EINECS: 233-171-7 UN: 2923 ADR: 8,II Merck Index: 12,10057 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...