Zirconium tetrafluoride 99% low hafnium Hf <0.1%
Supplier: Thermo Scientific Chemicals
|
Danger
|
11542-14
11542-22
CAAA11542-14EA
85.32
CAD
CAAA11542-14
CAAA11542-22
Zirconium tetrafluoride 99% low hafnium Hf <0.1%
Zirconium tetrafluoride
MDL: MFCD00011307
Purity: 99%
CAS No.: 7783-64-4
Molecular Formula: F4Zr
Formula Weight: 167.21
Physical Form: Powder
Density: 4.43
Refractive Index: 1.59
Sensitivity: Moisture Sensitive
Appearance: White
Melting Point: 600° subl.
Merck Reference: 14,10175
UN No.: 3260
MDL No.: MFCD00011307
Formula:
F₄Zr Melting Pt: 600 °C (subl.) Density: 4.43 Storage Temperature: Ambient |
MDL Number:
MFCD00011307 CAS Number: 7783-64-4 EINECS: 232-018-1 UN: 3260 ADR: 8,II Merck Index: 14,10175 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...