Titanium(IV) fluoride 98%

Supplier: Thermo Scientific Chemicals
Danger

11540-14 11540-22
CAAA11540-14EA 148.68 CAD
CAAA11540-14 CAAA11540-22
Titanium(IV) fluoride 98%
Titanium (IV) fluoride

MDL: MFCD00042545


Purity: 98% CAS No.: 7783-63-3 Molecular Formula: TiF4 Formula Weight: 123.89 Physical Form: Crystalline Density: 2.798 Appearance: White Odor: Odorless Melting Point: 284° subl. Merck Reference: 14,9479 UN No.: 3260 MDL No.: MFCD00042545

Formula: F₄Ti
MW: 123.87 g/mol
Boiling Pt: 284
Melting Pt: 400 (subl.)
Density: 2.798
Storage Temperature: Ambient
MDL Number: MFCD00042545
CAS Number: 7783-63-3
EINECS: 232-017-6
UN: 3260
ADR: 8 II
Merck Index: 14,9479

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR