Silver tetrafluoroborate 99%

Supplier: Thermo Scientific Chemicals
Danger

11539-09 11539-18
CAAA11539-09EA 205.4 CAD
CAAA11539-09 CAAA11539-18
Silver tetrafluoroborate 99%
Silver tetrafluoroborate

MDL: MFCD00003408 Fieser: 1,1015 4,428 5,587 6,519 8,443 9,414 11,471 18,322 21,394


Purity: 99% CAS No.: 14104-20-2 Molecular Formula: AgBF4 Formula Weight: 194.67 Physical Form: Crystalline Sensitivity: Light Sensitive and Hygroscopic Appearance: White Melting Point: 200° dec. UN No.: 3260 MDL No.: MFCD00003408

Formula: AgBF₄
Melting Pt: 200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003408
CAS Number: 14104-20-2
EINECS: 237-956-5
UN: 3260
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR