Silver hexafluoroantimonate 99%

Supplier: Thermo Scientific Chemicals
Danger

11538-03 11538-06 11538-14
CAAA11538-03EA 45.82 CAD
CAAA11538-03 CAAA11538-06 CAAA11538-14
Silver hexafluoroantimonate 99%
Silver hexafluoroantimonate

MDL: MFCD00003401 Fieser: 5,577 7,320 11,467


Purity: 99% CAS No.: 26042-64-8 Molecular Formula: AgSbF6 Formula Weight: 343.41 Physical Form: Powder/Lump Sensitivity: Light Sensitive and Hygroscopic Appearance: White to cream UN No.: 3260 MDL No.: MFCD00003401

Formula: AgF₆Sb
Density: 4,65…4.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003401
CAS Number: 26042-64-8
EINECS: 247-429-1
UN: 3260
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR