Tin (II) sulfate ≥95.5%

Supplier: Thermo Scientific Chemicals
Warning

11537-30 11537-A1
CAAA11537-30EA 115.5 CAD
CAAA11537-30 CAAA11537-A1
Tin (II) sulfate ≥95.5%
Tin (II) sulphate

Tin plating baths


Purity: 95.5% min CAS No.: 7488-55-3 Molecular Formula: SnSO4 Formula Weight: 214.77 Physical Form: Powder Density: 4.15 Melting Point: 378° dec. Solubility: Soluble in water and dilute H2SO4. Water solution hydrolyzes rapidly with precipitation of basic salt Merck Reference: 14,8790 MDL No.: MFCD00011246


Tin plating baths

Formula: O₄SSn
Melting Pt: 378 °C (dec.)
Density: 4.15
Storage Temperature: Ambient
MDL Number: MFCD00011246
CAS Number: 7488-55-3
EINECS: 231-302-2
Merck Index: 14,8790

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR