Tin(II) chloride dihydrate, REacton®, Reagent Grade

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Stannous chloride dihydrate

11536-22 11536-36 11536-A3
CAAA11536-22EA 68.26 CAD
CAAA11536-22 CAAA11536-36 CAAA11536-A3
Tin(II) chloride dihydrate, REacton®, Reagent Grade
Tin (II) chloride dihydrate

MDL: MFCD00149863 Soluble in HCl (dilute or concentrated), alcohol, ethyl acetate, glacial acetic acid, and NaOH solution. Decomposes in excess water. Fieser: 1,1113 15,309 16,329 17,334 18,353 19,337 20,373 21,422


CAS No.: 10025-69-1 Molecular Formula: SnCl2.2H2O Formula Weight: 225.65 (189.62anhy) Physical Form: Crystalline Density: 2.71 Sensitivity: Air Sensitive Solubility: Soluble in HCl (dilute or concentrated), alcohol, ethyl acetate, glacial acetic acid, and NaOH solution. Decomposes in excess water Merck Reference: 14,8783 UN No.: 3260 MDL No.: MFCD00149863

Formula: SnCl₂·2H₂O
MW: 225.65 g/mol
Boiling Pt: 623
Melting Pt: 246
Density: 2.71
Storage Temperature: Ambient
MDL Number: MFCD00149863
CAS Number: 10025-69-1
EINECS: 600-045-1
UN: 3260
ADR: 8 III
Merck Index: 14,8783

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR