Potassium hexafluorophosphate ≥95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
11531-30
11531-A1
CAAA11531-30EA
144.1
CAD
CAAA11531-30
CAAA11531-A1
Potassium hexafluorophosphate ≥95%
Potassium hexafluorophosphate
MDL: MFCD00011412
Purity: 95% min
CAS No.: 17084-13-8
Molecular Formula: KPF6
Formula Weight: 184.07
Physical Form: Crystalline
Density: 2.55
Appearance: White
Odor: Odorless
pH: 3.0-7.0
Melting Point: 575°
UN No.: 3260
MDL No.: MFCD00011412
Formula:
F₆KP Melting Pt: 575 °C Density: 2,55 Storage Temperature: Ambient |
MDL Number:
MFCD00011412 CAS Number: 17084-13-8 EINECS: 241-143-0 UN: 3260 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...