Lithium hexafluoroarsenate 99%

Supplier: Thermo Scientific Chemicals
Danger

11530-09 11530-18
CAAA11530-09EA 237 CAD
CAAA11530-09 CAAA11530-18
Lithium hexafluoroarsenate 99%
Lithium hexafluoroarsenate

MDL: MFCD00011076


Purity: 99% CAS No.: 29935-35-1 Molecular Formula: LiAsF6 Formula Weight: 195.85 Physical Form: Powder Sensitivity: Hygroscopic UN No.: 1557 MDL No.: MFCD00011076

Formula: AsF₆Li
MW: 195.85
Melting Pt: 349
Storage Temperature: Ambient
MDL Number: MFCD00011076
CAS Number: 29935-35-1
EINECS: 249-963-0
UN: 1557
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR