Lithium hexafluorophosphate 98%

Supplier: Thermo Scientific Chemicals
Danger

11529-03 11529-09 11529-18
CAAA11529-03EA 77.42 CAD
CAAA11529-03 CAAA11529-09 CAAA11529-18
Lithium hexafluorophosphate 98%
Lithium hexafluorophosphate

Used as an electrolyte in Li-ion batteries.


Purity: 98% CAS No.: 21324-40-3 Molecular Formula: LiPF6 Formula Weight: 151.90 Storage Temperature: 2°C to 8°C Physical Form: Powder Density: 1.5 Sensitivity: Moisture Sensitive Appearance: White Odor: Odorless Melting Point: 200° dec. UN No.: 2923 MDL No.: MFCD00011096


Used as an electrolyte in Li-ion batteries.

Formula: F₆LiP
Melting Pt: 200 °C (dec.)
Density: 1.5
Storage Temperature: Refrigerator
MDL Number: MFCD00011096
CAS Number: 21324-40-3
EINECS: 244-334-7
UN: 2923
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR