Lithium tetrafluoroborate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
11528-09
11528-18
CAAA11528-09EA
85.79
CAD
CAAA11528-09
CAAA11528-18
Lithium tetrafluoroborate 98%
Lithium tetrafluoroborate
MDL: MFCD00011087
Purity: 98%
CAS No.: 14283-07-9
Molecular Formula: BF4Li
Formula Weight: 93.75
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White to off-white
Odor: Odorless
Melting Point: 293-300° dec.
Merck Reference: 14,5543
UN No.: 3260
MDL No.: MFCD00011087
Formula:
BF₄Li MW: 93.74 g/mol Melting Pt: 293…300 °C (dec.) Density: 0.852 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011087 CAS Number: 14283-07-9 EINECS: 238-178-9 UN: 3260 ADR: 8,III Merck Index: 14,5543 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...