Manganese(II) fluoride 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Manganese(II) difluoride, Manganese difluoride
11527-09
11527-18
CAAA11527-09EA
66.68
CAD
CAAA11527-09
CAAA11527-18
Manganese(II) fluoride 99%
Manganese (II) fluoride
MDL: MFCD00016222
Insoluble in water, alcohol. Soluble in dilute HF, conc. HCl or HNO3
Purity: 99%
CAS No.: 7782-64-1
Molecular Formula: F2Mn
Formula Weight: 92.93
Physical Form: Crystalline Powder
Density: 3.98
Appearance: Red
Melting Point: 856°
Solubility: Insoluble in water, alcohol. Soluble in dilute HF, conc. HCl or HNO3
Merck Reference: 14,5729
UN No.: 3288
MDL No.: MFCD00016222
Formula:
F₂Mn MW: 92.93 Melting Pt: 856 °C Density: 3.98 Storage Temperature: Ambient |
MDL Number:
MFCD00016222 CAS Number: 7782-64-1 EINECS: 231-960-0 UN: 3288 ADR: 6.1 III Merck Index: 14,5729 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...