Platinum 99,99 Metals basis, foil, Premion®, Thickness: 0.025 mm (0.001 in)
Supplier: Thermo Scientific Chemicals
Synonyms:
Platinium
11510-FF
11510-FI
11510-GH
CAAA11510-FFEA
470.84
CAD
CAAA11510-FF
CAAA11510-FI
CAAA11510-GH
Platinum 99,99 Metals basis, foil, Premion®, Thickness: 0.025 mm (0.001 in)
Platinum
MDL: MFCD00011179
Purity: 99.99%
CAS No.: 7440-06-4
Physical Form: ≈0.34g/25x25mm
Density: 21.45G/CC
Odor: Odorless
MDL No.: MFCD00011179
Formula:
Pt MW: 195.08 g/mol Boiling Pt: 3827 Melting Pt: 1769 Density: 21.45G/CC Storage Temperature: Ambient |
MDL Number:
MFCD00011179 CAS Number: 7440-06-4 EINECS: 231-116-1 Merck Index: 12,07689 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...