Aluminium fluoride, anhydrous 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Aluminum fluoride

11497-03 11497-09 11497-18
CAAA11497-03EA 38.71 CAD
CAAA11497-03 CAAA11497-09 CAAA11497-18
Aluminium fluoride, anhydrous 99+%
Aluminium fluoride

In ceramics, as flux in metallurgy, in aluminum manufacture, as inhibitor of fermentation, as catalyst in organic reactions


Purity: 99+% CAS No.: 7784-18-1 Molecular Formula: AlF3 Formula Weight: 83.98 Physical Form: Powder Density: 2.882 Melting Point: 1291° subl. Solubility: Practically insoluble in water. Sparingly soluble in acids and alkalies Merck Reference: 14,339 MDL No.: MFCD00003426


In ceramics, as flux in metallurgy, in aluminum manufacture, as inhibitor of fermentation, as catalyst in organic reactions

Formula: AlF₃
Melting Pt: 1291 °C (subl.)
Density: 2.882
Storage Temperature: Ambient
MDL Number: MFCD00003426
CAS Number: 7784-18-1
EINECS: 232-051-1
UN: 3260
ADR: 8,II
Merck Index: 14,339

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR