Aluminium fluoride trihydrate ≥97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Aluminum fluoride trihydrate

11496-22 11496-36 11496-A3
CAAA11496-22EA 77.1 CAD
CAAA11496-22 CAAA11496-36 CAAA11496-A3
Aluminium fluoride trihydrate ≥97%
Aluminium fluoride trihydrate

MDL: MFCD00003426


Purity: 97% CAS No.: 15098-87-0 Molecular Formula: AlF3.3H2O Formula Weight: 138.02 (83.98anhy) Physical Form: Powder Density: 2.17 Appearance: White Odor: Odorless Melting Point: 250° -3H2O Merck Reference: 14,339 MDL No.: MFCD00003426

Formula: AlF₃·3H₂O
MW: 138.02 g/mol
Melting Pt: 250 °C (-3H₂O)
Density: 2.17
Storage Temperature: Ambient
MDL Number: MFCD00149141
CAS Number: 15098-87-0
EINECS: 232-051-1
UN: 3260
ADR: 8,III
Merck Index: 14,339

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR