Ammonium hexafluorophosphate ≥99.5%
Supplier: Thermo Scientific Chemicals
|
Danger
|
11495-14
11495-22
CAAA11495-14EA
105.54
CAD
CAAA11495-14
CAAA11495-22
Ammonium hexafluorophosphate ≥99.5%
Ammonium hexafluorophosphate
MDL: MFCD00064642
Freely soluble in water (75g/100ml). Soluble in acetone, methanol, ethanol, methyl acetate
Purity: 99.5%
CAS No.: 16941-11-0
Molecular Formula: NH4PF6
Formula Weight: 163.00
Physical Form: Powder
Density: 2.180
Appearance: Colorless to white
Melting Point: 198°
Solubility: Freely soluble in water (75g/100ml). Soluble in acetone, methanol, ethanol, methyl acetate
Merck Reference: 14,526
UN No.: 3260
MDL No.: MFCD00064642
Formula:
H₄F₆NP Melting Pt: 198 °C Density: 2.180 Storage Temperature: Ambient |
MDL Number:
MFCD00064642 CAS Number: 16941-11-0 EINECS: 241-009-1 UN: 3260 ADR: 8,III Merck Index: 14,526 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...