Antimony(III) fluoride 99+%

Supplier: Thermo Scientific Chemicals
Danger

11492-22 11492-36
CAAA11492-22EA 105.39 CAD
CAAA11492-22 CAAA11492-36
Antimony(III) fluoride 99+%
Antimony (III) fluoride

MDL: MFCD00011218 Fieser: 6,553


Purity: 99+% CAS No.: 7783-56-4 Molecular Formula: SbF3 Formula Weight: 178.75 Physical Form: Crystalline Density: 4.379 Melting Point: 292° Boiling Point: 376° Merck Reference: 14,709 UN No.: 2923 MDL No.: MFCD00011218

Formula: F₃Sb
Boiling Pt: 376 °C
Melting Pt: 292 °C
Density: 4.379
Storage Temperature: Ambient
MDL Number: MFCD00011218
CAS Number: 7783-56-4
EINECS: 232-009-2
UN: 2923
ADR: 8,III
Merck Index: 14,709

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR