Cobalt(III) fluoride 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cobaltic fluoride, Cobalt trifluoride

11490-14 11490-22
CAAA11490-14EA 145.68 CAD
CAAA11490-14 CAAA11490-22
Cobalt(III) fluoride 99%
Cobalt (III) fluoride

MDL: MFCD00010942


Purity: 99% CAS No.: 10026-18-3 Molecular Formula: CoF3 Formula Weight: 115.93 Physical Form: Powder Density: 3.88 Merck Reference: 14,2430 UN No.: 3085 MDL No.: MFCD00010942

Formula: CoF₃
MW: 115.93
Melting Pt: 927
Density: 3.88
Storage Temperature: Ambient
MDL Number: MFCD00010942
CAS Number: 10026-18-3
EINECS: 233-062-4
UN: 3085
ADR: 5.1 III
Merck Index: 14,2430

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR