Copper(II) fluoride, anhydrous ≥99.5%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Cupric fluoride
Cathodes in nonaqueous galvanic cells, high temp. fluorinating agent
Purity: 99.5%
CAS No.: 7789-19-7
Molecular Formula: CuF2
Formula Weight: 101.54
Physical Form: Powder
Density: 4.85
Appearance: White
Odor: Odorless
Melting Point: 950° dec.
Solubility: Solubility in water at 20° ca 4.7g/100ml
Merck Reference: 14,2638
UN No.: 3260
MDL No.: MFCD00010977
Cathodes in nonaqueous galvanic cells, high temp. fluorinating agent
Formula:
CuF₂ MW: 101.54 Boiling Pt: 950 Melting Pt: 855 (dec.) Density: 4.85 Storage Temperature: Ambient |
MDL Number:
MFCD00010977 CAS Number: 7789-19-7 EINECS: 232-147-3 UN: 3260 ADR: 8 III Merck Index: 14,2638 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...