Iron(III) fluoride, anhydrous ≥97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Iron trifluoride, Ferric fluoride

11487-09 11487-18
CAAA11487-09EA 66.99 CAD
CAAA11487-09 CAAA11487-18
Iron(III) fluoride, anhydrous ≥97%
Iron (III) fluoride

MDL: MFCD00011012 Slightly soluble in water. Freely soluble in dilute HF. Insoluble in benzene, ether, alcohol


Purity: 97% min CAS No.: 7783-50-8 Molecular Formula: FeF3 Formula Weight: 112.84 Physical Form: Powder Density: 3.87 Appearance: Pale green Melting Point: 1000° subl. Solubility: Slightly soluble in water. Freely soluble in dilute HF. Insoluble in benzene, ether, alcohol Merck Reference: 14,4022 UN No.: 3260 MDL No.: MFCD00011012

Formula: F₃Fe
MW: 112.84 g/mol
Melting Pt: >1000 °C (subl.)
Density: 3.87
Storage Temperature: Ambient
MDL Number: MFCD00011012
CAS Number: 7783-50-8
EINECS: 232-002-4
UN: 3260
ADR: 8 II
Merck Index: 14,4022

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR