Iron(II) fluoride, anhydrous 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ferrous fluoride, Iron difluoride
11486-09
11486-18
CAAA11486-09EA
132.09
CAD
CAAA11486-09
CAAA11486-18
Iron(II) fluoride, anhydrous 98%
Iron (II) fluoride
MDL: MFCD00016093
Purity: 98%
CAS No.: 7789-28-8
Molecular Formula: FeF2
Formula Weight: 93.84
Physical Form: Powder
Density: 4.09
Appearance: White
Melting Point:
Formula:
F₂Fe MW: 93.84 g/mol Boiling Pt: 1100 °C (1013 hPa) Melting Pt: >1000 °C (subl.) Density: 4.09 Storage Temperature: Ambient |
MDL Number:
MFCD00016093 CAS Number: 7789-28-8 EINECS: 232-155-7 UN: 3260 ADR: 8,II Merck Index: 14,4045 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...