Iron(II) iodide, anhydrous ≥97% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ferrous iodide, Iron diiodide
11485-04
11485-09
CAAA11485-04EA
125.29
CAD
CAAA11485-04
CAAA11485-09
Iron(II) iodide, anhydrous ≥97% (metals basis)
Iron (II) iodide
MDL: MFCD00016094
Purity: 97%
CAS No.: 7783-86-0
Molecular Formula: FeI2
Formula Weight: 309.66
Physical Form: Crystalline
Density: 5.32
Appearance: Brown
Odor: Odorless
UN No.: 3260
MDL No.: MFCD00016094
Formula:
FeI₂ MW: 309.66 g/mol Density: 5.32 Storage Temperature: Ambient |
MDL Number:
MFCD00016094 CAS Number: 7783-86-0 EINECS: 232-031-2 UN: 3260 ADR: 8,III Merck Index: 12,04097 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...